Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026125.3 | NP_001021296.1 | 7 - 8 | 1124 | 76 | |
NM_001380144.1 | NP_001367378.1 | 8 - 9 | 1157 | 76 | ||
NM_001380145.1 | NP_001367377.1 | 8 - 9 | 1265 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |