Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067940.5 | NP_500341.1 | 3 - 4 | 406 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |