Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307511.1 | NP_001294440.1 | 10 - 11 | 1379 | 76 | |
NM_001380150.1 | NP_001368270.1 | 11 - 12 | 1439 | 76 | ||
NM_067973.4 | NP_500374.2 | 6 - 7 | 991 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |