Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307511.1 | NP_001294440.1 | 12 - 13 | 1667 | 63 | |
NM_001380150.1 | NP_001368270.1 | 13 - 14 | 1727 | 63 | ||
NM_067973.4 | NP_500374.2 | 8 - 9 | 1279 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |