Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307511.1 | NP_001294440.1 | 13 - 14 | 1909 | 106 | |
NM_001380150.1 | NP_001368270.1 | 14 - 15 | 1969 | 106 | ||
NM_067973.4 | NP_500374.2 | 9 - 10 | 1521 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |