Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383068.1 | 2 - 3 | 564 | 84 | ||
GenBank mRNA | L19621.1 | - | 2 - 3 | 573 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |