Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxc-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068018.2 | NP_500419.1 | 2 - 3 | 238 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |