Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srz-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307706.2 | NP_001294635.1 | 4 - 5 | 768 | 142 | |
NM_001307707.3 | NP_001294636.1 | 2 - 3 | 436 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |