Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068038.4 | NP_500439.2 | 4 - 5 | 790 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |