Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068039.4 | NP_500440.2 | 2 - 3 | 501 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |