Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068048.6 | NP_500449.2 | 4 - 5 | 792 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |