Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028240.6 | NP_001023411.1 | 10 - 11 | 1443 | 69 | |
NM_001028242.6 | NP_001023413.1 | 8 - 9 | 1147 | 69 | ||
NM_001028243.3 | NP_001023414.1 | 8 - 9 | 1312 | 69 | ||
NM_001364041.3 | NP_001350991.1 | 9 - 10 | 1174 | 69 | ||
NM_001372787.3 | NP_001360026.1 | 8 - 9 | 1452 | 69 | ||
NM_001380173.1 | NP_001368701.1 | 10 - 11 | 1249 | 69 | ||
NM_001383072.1 | 10 - 11 | 1412 | 69 | |||
NM_001383073.1 | 10 - 11 | 1383 | 69 | |||
NM_068060.5 | NP_500461.1 | 9 - 10 | 1368 | 69 | ||
NM_068061.6 | NP_500462.1 | 9 - 10 | 1385 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |