Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068103.5 | NP_500504.1 | 2 - 3 | 269 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |