Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307635.3 | NP_001294564.1 | 2 - 3 | 329 | 71 | |
NM_068105.3 | NP_500506.2 | 3 - 4 | 494 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |