Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068129.3 | NP_500530.2 | 3 - 4 | 381 | 88 |
Gene Symbol: F15E6.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068130.5 | NP_500531.2 | 18 - 19 | 3931 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |