Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068154.7 | NP_500555.2 | 4 - 5 | 976 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |