Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wapl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372798.2 | NP_001360111.1 | 11 | 2498 | 65 | |
NM_001380189.1 | NP_001368334.1 | 1 - 2 | 234 | 65 | ||
NM_068165.6 | NP_500566.1 | 12 - 13 | 2172 | 65 | ||
NM_068166.8 | NP_500567.1 | 12 - 13 | 2166 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |