Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wapl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372798.2 | NP_001360111.1 | 4 | 572 | 58 | |
NM_001380190.1 | NP_001368333.1 | 4 - 5 | 578 | 58 | ||
NM_068165.6 | NP_500566.1 | 4 - 5 | 578 | 58 | ||
NM_068166.8 | NP_500567.1 | 4 - 5 | 572 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |