Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068178.5 | NP_500579.3 | 7 - 8 | 954 | 121 | |
GenBank mRNA | EU729335.1 | - | 7 - 8 | 815 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |