Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fem-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068423.6 | NP_500824.1 | 1 - 2 | 207 | 101 | |
NM_068424.4 | NP_500825.1 | 2 - 3 | 166 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |