Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fem-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068423.6 | NP_500824.1 | 8 - 9 | 1124 | 83 | |
NM_068424.4 | NP_500825.1 | 9 - 10 | 1090 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |