Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027894.7 | NP_001023065.1 | 4 - 5 | 1512 | 101 | |
NM_001380240.1 | NP_001367663.1 | 3 - 4 | 922 | 101 | ||
NM_068457.6 | NP_500858.2 | 4 - 5 | 1321 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |