Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068480.1 | NP_500881.1 | 3 - 4 | 426 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |