Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306838.3 | NP_001293767.1 | 4 - 5 | 669 | 74 | |
NM_068493.8 | NP_500894.1 | 6 - 7 | 1400 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |