Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380249.1 | NP_001367345.1 | 8 - 9 | 3018 | 68 | |
NM_001380252.2 | NP_001368407.1 | 8 - 9 | 3023 | 68 | ||
NM_001380254.1 | NP_001368409.1 | 3 - 4 | 519 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |