Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380250.2 | NP_001367343.1 | 7 - 8 | 3018 | 72 | |
NM_001380251.2 | NP_001367344.1 | 7 - 8 | 3015 | 72 | ||
NM_001380253.1 | NP_001368408.1 | 2 - 3 | 500 | 72 | ||
NM_068500.5 | NP_500901.2 | 1 - 2 | 287 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |