Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380249.1 | NP_001367345.1 | 15 - 16 | 5613 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |