Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268349.2 | NP_001255278.1 | 10 - 11 | 2802 | 107 | |
NM_001268350.4 | NP_001255279.2 | 11 - 12 | 2821 | 107 | ||
NM_001268351.3 | NP_001255280.1 | 10 - 11 | 3016 | 107 | ||
NM_001268352.3 | NP_001255281.1 | 10 - 11 | 2617 | 107 | ||
NM_001380258.1 | NP_001368424.1 | 8 - 9 | 2449 | 107 | ||
GenBank mRNA | FJ593901.1 | - | 10 - 11 | 2671 | 107 | |
FJ593903.1 | - | 10 - 11 | 2617 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |