Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268349.2 | NP_001255278.1 | 12 - 13 | 3092 | 73 | |
NM_001268350.4 | NP_001255279.2 | 13 - 14 | 3111 | 73 | ||
NM_001268351.3 | NP_001255280.1 | 12 - 13 | 3306 | 73 | ||
NM_001268352.3 | NP_001255281.1 | 12 - 13 | 2907 | 73 | ||
NM_001380258.1 | NP_001368424.1 | 10 - 11 | 2739 | 73 | ||
GenBank mRNA | FJ593901.1 | - | 12 - 13 | 2961 | 73 | |
FJ593903.1 | - | 12 - 13 | 2907 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |