Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068530.6 | NP_500931.3 | 3 - 4 | 462 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |