Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028211.4 | NP_001023382.1 | 8 - 9 | 1105 | 90 | |
NR_131543.1 | 8 - 9 | 1114 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |