Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smg-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380273.1 | NP_001368064.1 | 8 - 9 | 2567 | 86 | |
NM_068573.9 | NP_500974.2 | 8 - 9 | 2571 | 86 | ||
GenBank mRNA | EF472000.1 | - | 8 - 9 | 2585 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |