Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028406.5 | NP_001023577.1 | 8 - 9 | 1345 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |