Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wht-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069763.7 | NP_502164.1 | 11 - 12 | 1690 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |