Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rib-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069779.5 | NP_502180.1 | 5 - 6 | 569 | 129 | |
GenBank mRNA | AB241458.1 | - | 5 - 6 | 604 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |