Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tag-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380473.1 | NP_001366838.1 | 10 - 11 | 1608 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |