Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: let-92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383211.1 | 5 - 6 | 655 | 63 | ||
GenBank mRNA | AB108533.1 | - | 4 - 5 | 594 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |