Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: let-92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383211.1 | 6 - 7 | 822 | 92 | ||
GenBank mRNA | AB108533.1 | - | 5 - 6 | 761 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |