Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307047.3 | NP_001293976.1 | 2 - 3 | 331 | 56 | |
NM_069853.5 | NP_502254.1 | 2 - 3 | 263 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |