Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fat-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268666.3 | NP_001255595.1 | 5 - 6 | 798 | 115 | |
NM_001268667.3 | NP_001255596.1 | 3 - 4 | 613 | 115 | ||
NM_001268668.3 | NP_001255597.1 | 2 - 3 | 415 | 115 | ||
GenBank mRNA | AF260244.1 | - | 4 - 5 | 745 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |