Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tgt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069867.7 | NP_502268.2 | 2 - 3 | 1011 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |