Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268695.2 | NP_001255624.1 | 2 - 3 | 249 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |