Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268709.3 | NP_001255638.1 | 3 - 4 | 874 | 81 | |
NM_001268710.3 | NP_001255639.1 | 3 - 4 | 874 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |