Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: him-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069989.7 | NP_502390.2 | 4 - 5 | 1223 | 68 | |
GenBank mRNA | AY095296.1 | - | 4 - 5 | 1245 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |