Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ham-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070000.7 | NP_502401.1 | 4 - 5 | 992 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |