Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efa-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129346.3 | NP_001122818.1 | 4 - 5 | 661 | 78 | |
NM_001268723.3 | NP_001255652.1 | 2 - 3 | 263 | 78 | ||
NM_070015.6 | NP_502416.1 | 6 - 7 | 872 | 78 | ||
NM_070016.6 | NP_502417.1 | 6 - 7 | 872 | 78 | ||
GenBank mRNA | EF473217.1 | - | 5 - 6 | 815 | 78 | |
EF473218.1 | - | 4 - 5 | 656 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |