Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efa-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129346.3 | NP_001122818.1 | 2 - 3 | 308 | 63 | |
NM_070016.6 | NP_502417.1 | 3 - 4 | 366 | 63 | ||
GenBank mRNA | EF473218.1 | - | 2 - 3 | 303 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |