Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ark-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268725.3 | NP_001255654.1 | 11 - 12 | 3136 | 60 | |
GenBank mRNA | AJ271057.1 | - | 11 - 12 | 3136 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |