Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ark-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268724.2 | NP_001255653.1 | 5 - 6 | 952 | 63 | |
NM_001268725.3 | NP_001255654.1 | 5 - 6 | 952 | 63 | ||
GenBank mRNA | AJ271057.1 | - | 5 - 6 | 952 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |