Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ark-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268724.2 | NP_001255653.1 | 8 - 9 | 2196 | 75 | |
NM_001268725.3 | NP_001255654.1 | 8 - 9 | 2196 | 75 | ||
GenBank mRNA | AJ271057.1 | - | 8 - 9 | 2196 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |